Drucken
Anti-ZMYND11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-7697R-A555
BOSSBS-7697R-A555EA 893 EUR
BOSSBS-7697R-A555
Anti-ZMYND11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Chromatin reader that specifically recognizes and binds histone H3.3 trimethylated at 'Lys-36' (H3.3K36me3) and regulates RNA polymerase II elongation. Does not bind other histone H3 subtypes (H3.1 or H3.2) (By similarity). Colocalizes with highly expressed genes and functions as a transcription corepressor by modulating RNA polymerase II at the elongation stage. Acts as a tumor-suppressor by repressing a transcriptional program essential for tumor cell growth.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZMYND11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat