Drucken

Anti-Adenovirus 5 E1A binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-7697R-A750
BOSSBS-7697R-A750EA 893 EUR
BOSSBS-7697R-A750
Anti-Adenovirus 5 E1A binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Chromatin reader that specifically recognises and binds histone H3.3 trimethylated at 'Lys-36' (H3.3K36me3) and regulates RNA polymerase II elongation. Does not bind other histone H3 subtypes (H3.1 or H3.2) (By similarity). Colocalises with highly expressed genes and functions as a transcription corepressor by modulating RNA polymerase II at the elongation stage. Acts as a tumor-suppressor by repressing a transcriptional program essential for tumor cell growth.

Type: Primary
Antigen: Adenovirus 5 E1A binding protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-602/602
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR