Drucken

Anti-CKAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-7745R-A350
BOSSBS-7745R-A350EA 893 EUR
BOSSBS-7745R-A350
Anti-CKAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Possesses microtubule stabilizing properties. Involved in regulating aneuploidy, cell cycling, and cell death in a p53/TP53-dependent manner.Tissue specificity: Abundant in testis, thymus, and in tumor derived cell lines, while barely detectable in liver, prostate, and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CKAP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR