Drucken
Anti-CKAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-7745R-A488
BOSSBS-7745R-A488EA 893 EUR
BOSSBS-7745R-A488
Anti-CKAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Possesses microtubule stabilizing properties. Involved in regulating aneuploidy, cell cycling, and cell death in a p53/TP53-dependent manner.Tissue specificity: Abundant in testis, thymus, and in tumor derived cell lines, while barely detectable in liver, prostate, and kidney.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CKAP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now