Drucken

Anti-CCNYL1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-7889R-A750
BOSSBS-7889R-A750EA 893 EUR
BOSSBS-7889R-A750
Anti-CCNYL1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
CCNYL1 belongs to the cyclin family, Cyclin Y subfamily and contains 1 cyclin N terminal domain. The specific function of CCNYL1 is not yet known.

Type: Primary
Antigen: CCNYL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 68-150/359
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR