Drucken

Anti-COMMD4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-8037R-A750
BOSSBS-8037R-A750EA 893 EUR
BOSSBS-8037R-A750
Anti-COMMD4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes (PubMed:21778237). Down-regulates activation of NF-kappa-B.

Type: Primary
Antigen: COMMD4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/199
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR