Drucken

Anti-KLHL10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-8054R-A680
BOSSBS-8054R-A680EA 877 EUR
BOSSBS-8054R-A680
Anti-KLHL10 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
May be a substrate-specific adapter of a CUL3-based E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins during spermatogenesis.

Type: Primary
Antigen: KLHL10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/608
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR