Drucken

Anti-UBE2A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-8375R-A555
BOSSBS-8375R-A555EA 893 EUR
BOSSBS-8375R-A555
Anti-UBE2A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates. The first step requires the ATP-dependent activation of the Ub C-terminus and the assembly of multi-Ub chains by the Ub-activating enzyme known as the E1 component. The Ub chain is then conjugated to the Ub-conjugating enzyme (E2) to generate an intermediate Ub-E2 complex. The Ub-ligase (E3) then catalyzes the transfer of Ub from E2 to the appropriate protein substrate. UBE2A (Ubiquitin-conjugating enzyme E2 A) and UBE2B (Ubiquitin-conjugating enzyme E2 B) are both Ub-conjugating enzymes that are essential to postreplication repair of UV-damaged DNA. UBE2A and UBE2B are both nuclear and cell membrane proteins that have been found to interact with Rad18.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBE2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR