Drucken
Anti-RNF70 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-9223R-A350
BOSSBS-9223R-A350EA 893 EUR
BOSSBS-9223R-A350
Anti-RNF70 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome (By similarity). May be involved in protein sorting.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF70
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat