Drucken

Anti-RNF70 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9223R-A555
BOSSBS-9223R-A555EA 893 EUR
BOSSBS-9223R-A555
Anti-RNF70 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome (By similarity). May be involved in protein sorting.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF70
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR