Drucken

Anti-RNF14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9225R-A488
BOSSBS-9225R-A488EA 893 EUR
BOSSBS-9225R-A488
Anti-RNF14 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Might act as an E3 ubiquitin-protein ligase which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes and then transfers it to substrates, which could be nuclear proteins. Could play a role as a coactivator for androgen- and, to a lesser extent, progesterone-dependent transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF14
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR