Drucken

Anti-RNF123 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9249R-A750
BOSSBS-9249R-A750EA 893 EUR
BOSSBS-9249R-A750
Anti-RNF123 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Catalytic subunit of the KPC complex that acts as E3 ubiquitin-protein ligase. Required for poly-ubiquitination and proteasome-mediated degradation of CDKN1B during G1 phase of the cell cycle.

Type: Primary
Antigen: RNF123
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/1314
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR