Drucken

Anti-ZFP103 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9382R-A750
BOSSBS-9382R-A750EA 893 EUR
BOSSBS-9382R-A750
Anti-ZFP103 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Acts as an E2-dependent E3 ubiquitin-protein ligase, probably involved in the ER-associated protein degradation pathway.

Type: Primary
Antigen: ZFP103
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-230/685
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR