Drucken

Anti-ZFP103 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9382R-CY7
BOSSBS-9382R-CY7EA 900 EUR
BOSSBS-9382R-CY7
Anti-ZFP103 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Acts as an E2-dependent E3 ubiquitin-protein ligase, probably involved in the ER-associated protein degradation pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZFP103
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR