Drucken

Anti-PDE4A8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9719R-A555
BOSSBS-9719R-A555EA 893 EUR
BOSSBS-9719R-A555
Anti-PDE4A8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The PDE4A8 is a testis germ cell-specific PDE4A variant. There are two PDE4A variants identified in rat testis (PDE4A8 and an un-characterized 76 kDa PDE4A protein). Spermatogenesis is accompanied by expression of a varied repertoire of PDE4 enzymes at different stages of cell differentiation that presumably perform specialized functions. Confocal microscopic examination revealed the presence of PDE4A8 in primary and secondary spermatids but not in mature spermatozoa. PDE4A8 inhibition results in increased apoptosis of the germ cells accompanied with reduction in the number of mature spermatozoa.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDE4A8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR