Drucken

Anti-TCF9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-9812R-A680
BOSSBS-9812R-A680EA 1350 EUR
BOSSBS-9812R-A680
Anti-TCF9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GCF, also known as C2orf3 (chromosome 2 open reading frame 3), transcription factor 9 (TCF-9) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of -Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1400 genes and makes up approximately 8% of the human genome.

Type: Primary
Antigen: TCF9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-700/781
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR