Drucken

Anti-CD134/OX40 Polyclonal Antibody

Lieferant: BosterBio
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

A02495
BSBTA02495EA 552 EUR
BSBTA02495
Anti-CD134/OX40 Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for CD134/OX40 detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: CD134/OX40
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR