Drucken

PicoKine Human PD-L1/B7-H1; ELISA Kit

Lieferant: BOSTER BIOLOGICAL TECHNOLOGY
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

EK1449
BSBTEK1449EA 646 EUR
BSBTEK1449
PicoKine Human PD-L1/B7-H1; ELISA Kit
Tests ELISAs

Sandwich high sensitivity ELISA kit for quantitative detection of human PD-L1/B7-H1.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect Human Pd L1/CD274 with <12 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Pd L1/CD274 tissue specificity: Highly expressed in the heart, skeletal muscle, placenta and lung. Weakly expressed in the thymus, spleen, kidney and liver. Expressed on activated T- and B-cells, dendritic cells, keratinocytes and monocytes. .


96wells/kit, with removable strips.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR