Drucken

Anti-CD40 Rabbit Polyclonal Antibody

Lieferant: BosterBio
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PA1019
BSBTPA1019EA 552 EUR
BSBTPA1019
Anti-CD40 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 5(CD40) detection. Tested with WB in Mouse.

  • CD40 is a cell surface receptor that is expressed on the surface of all mature B cells and also expressed on monocytes, dendritic cells, and thymic epithelium. CD40 is a member of the tumor necrosis factor(TNF) receptor superfamily and is the receptor for CD40 ligand. CD40 ligand(CD40L, CD154, gp39, and TRAM) belongs to the TNF gene family and is expressed more widely than CD40 predominantly on activated CD4+ T cells. Activation of CD40 has also been shown to inhibit the growth of certain B cell lymphomas and to induce the death of transformed cells of mesenchymal or epithelial origin.

Each vial contains 50% glycerol, 0.9mg NaCl, 0.2mg Na2HPO4.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR