Drucken
Anti-BOB1 Rabbit Polyclonal Antibody
Lieferant: BosterBio
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
PA1506
BSBTPA1506EA 552 EUR
BSBTPA1506
Anti-BOB1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for POU domain class 2-associating factor 1(POU2AF1) detection. Tested with WB, IHC-P in Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: BOB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat