Drucken
Anti-CXCR5 Rabbit Polyclonal Antibody
Lieferant: BosterBio
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
PA2021
BSBTPA2021EA 552 EUR
BSBTPA2021
Anti-CXCR5 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for C-X-C chemokine receptor type 5 (CXCR5) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CXCR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat