Drucken

Anti-MCL1 Polyclonal Antibody

Lieferant: BosterBio
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PB9132
BSBTPB9132EA 552 EUR
BSBTPB9132
Anti-MCL1 Polyclonal Antibody
Antikörper
Polyclonal antibody for Mcl-1/MCL1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Mcl-1/MCL1 information: Molecular Weight: 37337 MW; Subcellular Localization: Membrane ; Single-pass membrane protein . Cytoplasm. Mitochondrion. Nucleus, nucleoplasm. Cytoplasmic, associated with mitochondria.

Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR