Drucken

Anti-DHFR/Dihydrofolate Reductase Polyclonal Antibody

Lieferant: BosterBio
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PB9175
BSBTPB9175EA 552 EUR
BSBTPB9175
Anti-DHFR/Dihydrofolate Reductase Polyclonal Antibody
Antikörper
Polyclonal antibody for Dihydrofolate Reductase/DHFR detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Dihydrofolate Reductase/DHFR information: Molecular Weight: 21453 MW; Tissue Specificity: Widely expressed in fetal and adult tissues, including throughout the fetal and adult brains and whole blood. Expression is higher in the adult brain than in the fetal brain.

Type: Primary
Antigen: DHFR/Dihydrofolate Reductase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR