Drucken

Anti-Kallikrein 4 Polyclonal Antibody

Lieferant: BosterBio
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PB9260
BSBTPB9260EA 552 EUR
BSBTPB9260
Anti-Kallikrein 4 Polyclonal Antibody
Antikörper
Polyclonal antibody for Kallikrein 4/KLK4 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Kallikrein 4/KLK4 information: Molecular Weight: 27032 MW; Subcellular Localization: Secreted; Tissue Specificity: Expressed in prostate.

Type: Primary
Antigen: Kallikrein 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR