Drucken

Anti-Stefin B Rabbit Polyclonal Antibody

Lieferant: BosterBio
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PB9565
BSBTPB9565EA 552 EUR
BSBTPB9565
Anti-Stefin B Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cystatin-B(CSTB) detection. Tested with WB, IHC-P, ELISA in Human.

Type: Primary
Antigen: Stefin B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR