Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: rat
Lieferant: BIOSENSIS PTY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BEK-2230-1P BEK-2230-2P
BSENBEK-2230-1PEA 398 EUR InStock
BSENBEK-2230-1P BSENBEK-2230-2P
Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: rat
Tests ELISAs
The Biosensis GDNF RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the quantification of GDNF in less than 4 hours in cell culture supernatants, cell lysates, and serum only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application, in particular rat serum samples.
This ELISA kit consists of a pre-coated anti-GDNF capture antibody, a biotinylated anti-GDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’- tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of GDNF present in samples and protein standards. A GDNF positive control (QC sample) is provided to assure consistent assay performance.
This GDNF ELISA kit employs a recombinant rat GDNF standard and is therefore designed to measure the rat form of GDNF. Note that the antibodies used in this kit cross-react with mouse, human and guinea pig GDNF. This kit has not been tested for other applications.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular rat serum samples.
Lieferumfang: The ELISA kit box contains 1 or 2×96-well pre-coated strip plate, protein standards, QC sample, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Achtung! Bitte beachten: It has been configured for research use only and is not to be used in diagnostic or clinical procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge