Nerve growth factor receptor (NGFR/P75ECD) Rapid™ ELISA kit: Human
Lieferant: BIOSENSIS PTY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BEK-2239-1P BEK-2239-2P
BSENBEK-2239-1PEA 398 EUR InStock
BSENBEK-2239-1P BSENBEK-2239-2P
Nerve growth factor receptor (NGFR/P75ECD) Rapid™ ELISA kit: Human
Tests ELISAs
The Biosensis NGFR/p75ECD Rapid™ enzyme-linked immunosorbent assay (ELISA) kit is a sandwich ELISA that allows the quantification of the extracellular domain (ECD) of human p75 neurotrophin receptor in less than four hours in urine only if used as directed.
  • Complete kit with precise instructions
This optimised human p75ECD ELISA assay demonstrates an increased sensitivity, lowering the LOD about 8-fold to the previous assay version. It represents the most sensitive NGFR/p75ECD ELISA assay available today, with a detection limit of at least 4 pg/ml, and a quantification range of 7,8 to 500 pg/ml.
This ELISA kit consists of a pre-coated mouse monoclonal anti-p75ECD capture antibody, a biotinylated mouse monoclonal anti-p75ECD detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3',5,5'-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the concentration of p75ECD present in samples and protein standards. A human p75ECD positive control (QC sample) is provided to assure consistent assay performance.
This NGFR/p75ECD ELISA kit employs an in-house recombinant human p75ECD-Fc chimera as standard that has been validated against an externally available p75ECD-Fc protein. While there is a current lack of a commercially available, true human p75ECD standard, this ELISA kit allows quantification of human p75ECD as p75ECD-Fc human equivalents.
The new monoclonal antibody pair yields higher OD values in urine samples, demonstrating its increased ability to detect natural p75ECD in biological samples. This means that samples assayed with BEK-2239 will give higher urine p75ECD levels, or that urine samples from healthy controls with previous urine p75ECD concentrations
Lieferumfang: Each kit contains 1 or 2×96 pre-coated strip plates, protein standards, QC sample, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Achtung! Bitte beachten: For research use only. Not for diagnostic or clinical purposes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge