Drucken

Anti-PGR Mouse Monoclonal Antibody [clone: 608]

Lieferant: Cayman Chemical
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

10009763-1
CAYM10009763-1EA 645 EUR
CAYM10009763-1
Anti-PGR Mouse Monoclonal Antibody [clone: 608]
Antikörper
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 608
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR