Drucken

Superoxide dismutase assay kit

Lieferant: CAYMAN
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

706002-96 706002-480
CAYM706002-96EA 349 EUR
CAYM706002-96 CAYM706002-480
Superoxide dismutase assay kit
Tests Enzymtests

Superoxide Dismutase Assay (SOD) kit is a fast and reliable assay for the measurement of SOD activity from plasma, serum, tissue homogenates and cell lysates.


  • Measure copper/zinc, manganese, and iron SOD in tissues, plasma, serum, and cell lysates
  • Measure SOD activity down to 0,005 U/ml


Significant amounts of superoxide dismutase (SOD) in cellular and extracellular environments are crucial for the prevention of diseases linked to oxidative stress. Mutations in SOD account for approximately 20% of familial amyotrophic lateral sclerosis (ALS) cases. SOD also appears to be important in the prevention of other neurodegenerative disorders such as Alzheimer’s, Parkinson’s, and Huntington’s Diseases. The reaction catalysed by SOD is extremely fast, having a turnover of 2×10⁶ M⁻¹sec⁻¹ and the presence of sufficient amounts of the enzyme in cells and tissues typically keeps the concentration of superoxide very low. Quantification of SOD activity is therefore essential in order to fully characterise the antioxidant capabilities of a biological system.


Each kit contains sufficient reagents to assay 41 samples in duplicate and includes assay buffer, sample buffer, radical detector, SOD (standard), xanthine oxidase, a 96 well plate, and complete instructions.


Lieferumfang: Kit contains sufficient reagents to assay 41 samples in duplicate and includes assay buffer, sample buffer, radical detector, SOD (standard), xanthine oxidase, a 96 well solid plate (colorimetric assay) and complete instructions.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR