Drucken

(-)-Epigallocatechin gallate ≥98%, kristalliner Feststoff

Lieferant: CAYMAN
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

70935-100 70935-50 70935-500 70935-250
CAYM70935-100EA 91 EUR
CAYM70935-100 CAYM70935-50 CAYM70935-500 CAYM70935-250
(-)-Epigallocatechin gallate ≥98%, kristalliner Feststoff
(-)-Epigallocatechin gallate

(–)-Epigallocatechin gallate (EGCG) is a phenol, found in a variety of plants including green and black tea plants and has diverse biological activities.

Formel: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00075940
CAS-Nummer: 989-51-5

Order Now

Spezifikation Testergebnisse

Purity ≥98%
λmax 276 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR