Drucken
Phoenix BLACK™ Sulfated Ash Muffle Furnace
Lieferant: CEM
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
928254-5
CEMA928254-5EA 0 EUR
CEMA928254-5
Phoenix BLACK™ Sulfated Ash Muffle Furnace
Öfen Veraschungs- und Muffelöfen
The Phoenix BLACK™ muffle furnace utilises microwave energy to quickly reach temperature and maintain 2,8 m³ (100 ft³) of air flow per minute through the cavity to rapidly ash materials and remove combustion by-products. Unit comes configured with the Sulfated ash option that allows for easy installation of all glassware and provides connections for needed accessories. Manufactured by CEM® Corporation, ISO-9001 certified. This technology is protected under US patent 5,318,754.
Verpackung: Includes:
Furnace - High capacity furnace insert for use in the Phoenix BLACK™ Sulfated ash, safe to 1000 °C. Holds up to (15) 20/50 ml CEM® Quartz Fiber Crucibles or (12) 100 ml CEM® Quartz fiber crucibles. Includes insert, flue, heating element, quartz plate/ceiling, chimney, and dual thermocouple.
Accessory kit - Contains furnace insert cooling fan, scrubber assembly, 243,84 cm exhaust hose, applicable fuses, and all necessary glassware to perform Sulfated Ash in the Phoenix BLACK™ as well as (4) 50 ml CEM® Quartz Fiber Crucibles, (4) 20 ml CEM® Quartz Fiber Crucibles, (1) crucible pen and operation manual.
Vacuum Pump, 50 Hz - Used to draw acid vapors through Phoenix BLACK™ Sulfated Ash scrubber system.0
Appropriate power cord(s) and adapters