Drucken
Needle-Free Fluid Transfer System, CODAN Spikes / MicroSpikes
Lieferant: CODAN
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
165260 16.5292 16.5264 165274 79.3800 79.3801
CODA165260EA 3770 EUR
CODA165260 CODA16.5292 CODA16.5264 CODA165274 CODA79.3800 CODA79.3801
Needle-Free Fluid Transfer System, CODAN Spikes / MicroSpikes
Kanülen
The CODAN Spikes are a compact and safe needle-free system for the manual preparation, withdrawal or transfer of cytostatics and pharmaceuticals as well as their solvent solutions. In order to protect users from aerosols and the drug from micro bacterial contamination, all CODAN Spikes are equipped with a hydrophobic 0,2 µm air filter.
  • Gamma sterilised
  • Cap protects against damage and contamination
  • Integrated, hydrophobic 0,2 µm air filter guarantees sterile aeration and venting and reliably retains aerosols during preparation
  • Minimise the risk of injury
  • Maximise the operating speed by work simplification
The CODAN MicroSpikes were especially designed for use on very small medication vials, e.g. preparation of vaccine.
For the filtration of large particles (e.g. plug punching, glass splinters) CODAN offers Chemoprotect® Spikes with an additional 5 µm fluid filter.
CODAN Spikes / MicroSpikes are also available with SWAN-LOCK® (SWabable Adapter Needle-free - Luer-Lock) access for multi-dose applications.
The handy Chemoprotect® bag with a safe-seal closure is ideal for the transport of medication to the ward and is also excellent for the disposal of substances.