Drucken
Thermal printer
Lieferant: CONSORT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
AP414
CONSAP414EA 0 EUR
CONSAP414
Thermal printer
Drucker Document Printers
This thermal printer can deliver both serial and parallel input when using RS232 and Centronix connectors respectively. Direct connection is available to connect all meters with digital output.
  • Prints 40 columns in normal and 80 columns in condensed condition
  • Roll length: ±28 m
Lieferumfang: Supplied with manual, roll of thermal paper, mains adaptor (230 VAC) and RS232 cable.