Drucken
Cleaning swabs, sealed foam, CONSTIX® SF-1
Lieferant: CONTEC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
SF-1
CONTSF-1EA 272 EUR
CONTSF-1
Cleaning swabs, sealed foam, CONSTIX® SF-1
Tupfer Reinigungstupfer
The SF-1 sealed foam swab consists of polyurethane foam bonded to a PP handle without the use of adhesives or binders.
  • Polyurethane foam head provides good particle entrapment and removal
  • Large head is ideal for cleaning and applying coating
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal.
Though compatible with many common solvents, the SF-12 is not recommended for use with ketones such as acetone or MEK.
Verpackung: Swabs are packed 500 per case.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904