Drucken
Flocked swabs
Lieferant: COPAN
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
961C 961CS01
COPA961CEA 889 EUR
COPA961C COPA961CS01
Flocked swabs
Tupfer Tupfer für die Probenahme
An extensive range of ready-to-use environmental swabs. The swabls are designed for use in a broad range of environments, including food production, pharmaceutical sites, as well as hospital wards, to determine the microbial bioburden.
  • Sterile options available
  • Varied packaging options
The nylon-flocked swab is designed to improve sample absorption by strong capillary action and to release more than 80% of the collected sample. This increases the efficiency in detecting microorganisms. Traditionally wrapped fiber swabs (rayon or polyester) trap in the fiber matrix a large percentage of the sample, releasing only 20% of it.