Drucken
Büchner-Trichter, PYREX®
Lieferant: Corning
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
36060-30C
CORN36060-30CEA 0 EUR
CORN36060-30C
Büchner-Trichter, PYREX®
Trichter Filtertrichter Büchner-Trichter
PYREX®, borosilicate glass, with sintered glass disc.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Especially useful where filter paper would be attacked by material being filtered. Top edge is beaded.
Achtung! Bitte beachten: Disc should not be subjected to pressure differential in excess of 1,01 bar.