Drucken
Modified Impeller Assembly for Magnetically-Driven Corning® Proculture® Spinner Flask
Lieferant: Corning
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
401392 401661 401901
734-1800EA 216.96 EUR
734-1800 734-1801 CORN401901
Modified Impeller Assembly for Magnetically-Driven Corning® Proculture® Spinner Flask
Kolben Spinnerkolben
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, Proculture® Spinner Flasks
  • Narrower impeller paddle design allows use with probes to create a small bioreactor
  • Non sterile