Drucken
Bottle Carrier
Lieferant: DELTALAB
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
19700
DELT19700EA 229 EUR
DELT19700
Bottle Carrier
Transportbehälter Flaschentragebehälter
Bottle carrier made of stainless steel, with folding handles.
  • Very good chemical resistance
  • Very good temperature resistance
It is suitable for the transport of up to six (1 L) chemical reagent bottles.