Drucken
Drums, wide neck, with screw cap and tap
Lieferant: DELTALAB
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
209030
DELT209030EA 105 EUR
DELT209030
Drums, wide neck, with screw cap and tap
Fässer, Fässer, Trommeln
PE, wide neck, with green PE screw cap.
  • Cap with non hermetical plug
  • With a tap and two black folding handles at the top
  • Not designed to transport liquids