Drucken
Flaschen zur Wasserprobenahme, rechteckig
Lieferant: DELTALAB
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
282331 282330 282320 282321 282323 282323.BU 282324 282324.BU
DELT282331EA 94.1 EUR
DELT282331 DELT282330 DELT282320 DELT282321 DELT282323 DELT282323.BU DELT282324 DELT282324.BU
Flaschen zur Wasserprobenahme, rechteckig
Flaschen Flaschen zur Wasserprobenahme
Polyethylene, translucent, with red screw cap.
  • Tamper evident red cap with an internal joint
  • Hermetical closure
  • Sterilised by radiation
  • Available with or without sodium thiosulfate
With liquid sodium thiosulphate (24 mg/L): ideal to analyse water meant for human consumption, swimming pools, and any other water where chlorine may modify the composition of the sample while being transported.
Without sodium thiosulphate: ideal to sample water for physico-chemical analysis, as well as for other sampling requiring sterile bottles.
The minor sides are gripped for a better handling. The larger sides are flat for labelling. Each bottle is labelled specifying description, code, lot, and expiry date.