Drucken
Dri-Shield® 3000 Moisture Barrier Bag
Lieferant: DESCO
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
D301216
DESCD301216EA 139 EUR
DESCD301216
Dri-Shield® 3000 Moisture Barrier Bag
Beutel Aufbewahrungsbeutel
This foil moisture barrier bag provides protection for ESD sensitive and moisture sensitive electronics, typically SMT (Surface Mount Technology) items.
  • A nylon layer helps to strengthen the bag
  • The bags are heat sealable and suitable for vacuum packaging
  • Bags are printed with ESD protective and moisture sensitive warning symbols, and a lot code for traceability
SCS Moisture Barrier Bags Dri-Shield® 3000 are manufactured from a laminate of nylon, foil, and polyethylene. The metal layer provides discharge shielding and minimises the penetration of electric fields and electrostatic discharges.
Zertifizierungen: Meets ANSI/ESD S20.20, ANSI/ESD S541, ANSI/ESD S11.4 Level 1, and IPC/JEDEC J-STD-033.