Drucken
Digital Timer and Stopwatch, SHAKE AWAKE
Lieferant: TFA DOSTMANN
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
38.2019
DOST38.2019EA 28.9 EUR
DOST38.2019
Digital Timer and Stopwatch, SHAKE AWAKE
Timer Timer, digital
Wall mounting or table stand digital timer and stopwatch, SHAKE AWAKE.
  • With beep sound, flashing light or vibration alarm
  • Hours minutes and minutes/seconds display
  • With mounting magnet and clip
  • Ideal in noisy environments or for someone with hearing difficulties
Lieferumfang: Supplied with timer with stopwatch, batteries (2×AAA, 1,5 V), instruction manual.