Drucken
Genes in a Tube™
Lieferant: EDVOTEK
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
119
EDVO119EA 153 EUR
EDVO119
Genes in a Tube™
Lehrmittel Biologie-Lehrmittel Genetics Learning Activities
Kit for teaching students how to extract and precipitate their own DNA. Students can transfer their DNA to a tube that can be used as a pendant on a necklace.
  • For 25 students, plus 1
  • Complete in 30 minutes
Also required: Ice cold alcohol (95% ethanol or isopropyl alcohol) and a 56 °C water bath.
Also required: Ice cold alcohol (95% ethanol or isopropyl alcohol) and a 56 °C water bath.
Lieferumfang: Kit includes instructions, lysis buffer, NaCl solution, Protease, Tris buffer, Methylene Blue Plus solution, microcentrifuge tubes, sterile cotton tipped applicators, transfer pipettes, tubes for DNA precipitation, Gene Tubes, and string.