Drucken
Ready-to-Load™ kits, in search of my father
Lieferant: EDVOTEK
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
S-49
EDVOS-49EA 71.5 EUR
EDVOS-49
Ready-to-Load™ kits, in search of my father
Lehrmittel Biologie-Lehrmittel Genetics Learning Activities
Solve the mystery of two boys separated from their parents a decade ago. Their biological mother is identified by mitrochondrial DNA and their biological father from chromosomal DNA.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Lieferumfang: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.