Drucken
Restriction enzymes, NdeI, Fermentas
Lieferant: FERMENTAS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ER0581 ER0582 ER0585
ER0581EA 40.62 EUR
ER0581 ER0582 ER0585
Restriction enzymes, NdeI, Fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
NdeI restriction enzyme recognises CA^TATG sites and cuts at 37 °C in O buffer (Isoschizomers: FauNDI).
  • Used for molecular cloning, restriction site mapping, genotyping, southern blotting, RFLP and SNP.
Superior quality, stringent quality control and industry leading manufacturing process
Convenient colour-coded five buffer system
BSA premixed in reaction buffers
Wide selection of restriction endonuclease specificities
Lieferumfang: Supplied with universal Tango buffer for double-digestions.