Restriction enzymes, PfoI, Fermentas
Lieferant: FERMENTASGesamtbewertung: 0
Bewertung: von 5
PfoI restriction enzyme recognises T^CCNGGA sites and cuts best at 37 °C in Tango buffer.
- Stringent quality control and industry leading manufacturing process
- Colour-coded five buffer system
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Its application includes molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP)and SNP.
PdoI is CpG methylation-sensitive, dam methylation-sensitive, dcm methylation-sensitive and also sensitive to heat inactivation.
Lieferumfang: It includes universal Tango buffer for double-digestions.