Drucken

Anti-ATG4A Goat Polyclonal Antibody

Lieferant: Everest Biotech
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

EB08867
EVBIEB08867EA 339 EUR
EVBIEB08867
Anti-ATG4A Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR