Drucken

Anti-FGF5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

EB11181
EVBIEB11181EA 339 EUR
EVBIEB11181
Anti-FGF5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGF5 (fibroblast growth factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR