Drucken

Anti-MEX3C Goat Polyclonal Antibody

Lieferant: Everest Biotech
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

EB11536
EVBIEB11536EA 339 EUR
EVBIEB11536
Anti-MEX3C Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MEX3C (mex-3 RNA binding family member C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR