Drucken

Anti-BMP1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

EB12632
EVBIEB12632EA 339 EUR
EVBIEB12632
Anti-BMP1 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 110kDa band observed in Human Heart and Human Kidney lysates (calculated MW of 111kDa according to NP_006120.1). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: BMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR